Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016582.2 | NP_057666.1 | 6, 7 | 1675 | ||
NR_027391.1 | 5, 6 | 1506 | ||||
XM_011545095.2 | XP_011543397.1 | 6, 7 | 1970 | |||
GenBank mRNA | AB020598.1 | - | 6, 7 | 1675 | ||
AF135600.1 | - | 6, 7 | 1568 | |||
AK127216.1 | - | 1, 2 | 4130 | |||
BC025710.1 | - | 5, 6 | 1366 | 5224878 | ||
BC037974.1 | - | 6, 7 | 1535 | 5213632 | ||
BU731752.1 | - | 2, 3 | 456 |