Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271803.1 | NP_001258732.1 | 2 | 260 | ||
NM_016606.3 | NP_057690.2 | 2 | 260 | |||
NR_073448.1 | 2 | 335 | ||||
NR_073449.1 | 2 | 335 | ||||
GenBank mRNA | AF251041.1 | - | 1 | 33 | ||
AK056193.1 | - | 2 | 216 | |||
AK223611.1 | - | 2 | 201 | |||
AK290547.1 | - | 2 | 150 | |||
AK293480.1 | - | 2 | 234 | |||
AY562240.1 | - | 2 | 52 | |||
BC006218.2 | - | 2 | 174 | 3940709 | ||
DC374786.1 | - | 2 | 260 | |||
DC424086.1 | - | 2 | 286 |