Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYND10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308379.1 | NP_001295308.1 | 8 | 1101 | ||
NM_015896.3 | NP_056980.2 | 9 | 1116 | |||
XM_005265216.2 | XP_005265273.1 | 8 | 1289 | |||
GenBank mRNA | AB209621.1 | - | 1 | 4462 | ||
AK058135.1 | - | 9 | 1046 | |||
AK093455.1 | - | 1 | 1104 | |||
AK096525.1 | - | 9 | 2224 | |||
AK223343.1 | - | 9 | 1031 | |||
AK308086.1 | - | 9 | 1013 | |||
AK315123.1 | - | 8 | 1087 | |||
AL833828.1 | - | 9 | 1045 | |||
BC033732.1 | - | 9 | 1012 | 5168611 | ||
U70824.1 | - | 9 | 1070 | |||
U70880.1 | - | 8 | 1055 |