Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RWDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007464.2 | NP_001007465.1 | 4 | 566 | ||
NM_015952.3 | NP_057036.2 | 3 | 452 | |||
NM_016104.3 | NP_057188.2 | 5 | 694 | |||
GenBank mRNA | AF092134.1 | - | 3 | 409 | ||
AF132958.1 | - | 3 | 322 | |||
AK290727.1 | - | 3 | 452 | |||
BC015802.1 | - | 3 | 308 | 3909348 | ||
CA422968.1 | - | 5 | 675 | |||
CF128570.1 | - | 3 | 399 | |||
CN363729.1 | - | 5 | 535 |