Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016255.2 | NP_057339.1 | 1, 2 | 837 | ||
GenBank mRNA | AF097027.1 | - | 1, 2 | 769 | ||
AK296393.1 | - | 2, 3 | 151 | |||
AK312814.1 | - | 1, 2 | 772 | |||
BC047881.1 | - | 1, 2 | 761 | 5742372 |