Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172828.1 | NP_001166299.1 | 3 | 1721 | ||
NM_001173128.1 | NP_001166599.1 | 5 | 1828 | |||
NM_016258.2 | NP_057342.2 | 4 | 2062 | |||
GenBank mRNA | AB451335.1 | - | 4 | 1565 | ||
AB451484.1 | - | 4 | 1565 | |||
AF155095.1 | - | 4 | 1715 | |||
AF192968.1 | - | 4 | 1715 | |||
AF432214.1 | - | 3 | 2245 | |||
AK292581.1 | - | 4 | 1864 | |||
AK303833.1 | - | 3 | 1721 | |||
BC002559.2 | - | 4 | 1927 | 3139250 |