Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006204.3 | NP_006195.3 | 4 | 975 | ||
XM_017016322.1 | XP_016871811.1 | 2 | 339 | |||
GenBank mRNA | U31973.1 | - | 4 | 975 |