Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002602.3 | NP_002593.1 | 4 | 351 | ||
NR_026872.1 | 3 | 146 | ||||
XM_006721942.3 | XP_006722005.1 | 4 | 1324 | |||
XM_017024734.1 | XP_016880223.1 | 4 | 626 | |||
XM_017024735.1 | XP_016880224.1 | 4 | 357 | |||
XM_017024736.1 | XP_016880225.1 | 4 | 291 | |||
GenBank mRNA | BC106884.2 | - | 4 | 308 | 40029486 | |
BQ638010.1 | - | 4 | 292 | |||
BX537414.1 | - | 4 | 358 | |||
BX647779.1 | - | 3 | 140 | |||
M36476.1 | - | 4 | 308 |