Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3HC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282190.1 | NP_001269119.1 | 10 | 1458 | ||
NM_001282191.1 | NP_001269120.1 | 8 | 1199 | |||
NM_016478.4 | NP_057562.3 | 9 | 1412 | |||
XM_005250403.2 | XP_005250460.1 | 7 | 1072 | |||
XM_011516288.2 | XP_011514590.1 | 8 | 1238 | |||
XM_011516289.2 | XP_011514591.1 | 8 | 1623 | |||
XM_011516290.2 | XP_011514592.1 | 7 | 2962 | |||
XM_017012287.1 | XP_016867776.1 | 9 | 1278 | |||
XM_017012288.1 | XP_016867777.1 | 6 | 1914 | |||
GenBank mRNA | AF151050.1 | - | 8 | 1120 | ||
AJ537494.1 | - | 9 | 1327 | |||
AK001317.1 | - | 10 | 1381 | |||
AK022373.1 | - | 9 | 1335 | |||
BC000190.1 | - | 9 | 1320 | |||
BC011551.1 | - | 9 | 1328 | 4561164 | ||
BC028917.1 | - | 4 | 604 |