Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016481.4 | NP_057565.3 | 4 | 723 | ||
XM_005252047.3 | XP_005252104.1 | 4 | 680 | |||
XM_005252048.4 | XP_005252105.1 | 5 | 783 | |||
XM_005252049.4 | XP_005252106.1 | 3 | 505 | |||
XM_005252050.4 | XP_005252107.1 | 4 | 609 | |||
XM_011518776.2 | XP_011517078.1 | 4 | 680 | |||
XM_011518777.1 | XP_011517079.1 | 5 | 762 | |||
GenBank mRNA | AF151053.1 | - | 4 | 668 | ||
AK000213.1 | - | 4 | 723 | |||
AK023069.1 | - | 1 | 2092 | |||
AK309270.1 | - | 5 | 835 | |||
AY189280.1 | - | 6 | 2777 | |||
BC002863.2 | - | 4 | 703 | 3944148 |