Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016538.2 | NP_057622.1 | 7 | 833 | ||
XM_006722284.2 | XP_006722347.1 | 5 | 567 | |||
XM_011523580.1 | XP_011521882.1 | 6 | 699 | |||
XM_011523581.2 | XP_011521883.1 | 6 | 2436 | |||
GenBank mRNA | AF233395.1 | - | 7 | 815 | ||
AK002027.1 | - | 5 | 1613 | |||
AK094326.1 | - | 6 | 898 | |||
AK131437.1 | - | 4 | 1890 | |||
AK131484.1 | - | 2 | 1388 | |||
AK290265.1 | - | 7 | 801 | |||
AL137626.1 | - | 4 | 401 | |||
BC017305.2 | - | 7 | 797 | 5087554 | ||
BC101791.1 | - | 7 | 814 | 8069297 | ||
BC101793.1 | - | 7 | 813 | 8069299 |