Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286703.1 | NP_001273632.1 | 5, 6 | 324 | ||
NM_001286704.1 | NP_001273633.1 | 5, 6 | 502 | |||
NM_016617.3 | NP_057701.1 | 5, 6 | 330 | |||
NR_104584.1 | 6, 7 | 391 | ||||
NR_104585.1 | 4, 5 | 290 | ||||
GenBank mRNA | AB154404.1 | - | 4, 5 | 181 | ||
AF208844.1 | - | 5, 6 | 220 | |||
AK095490.1 | - | 5, 6 | 250 | |||
AL515797.3 | - | 4, 5 | 182 | |||
BC005193.1 | - | 5, 6 | 247 | 3829206 | ||
BI458294.1 | - | 5, 6 | 301 | |||
BI761948.1 | - | 5, 6 | 207 | |||
BP277186.1 | - | 5, 6 | 330 | |||
BU589105.1 | - | 6, 7 | 307 | |||
CR457189.1 | - | 4, 5 | 181 | |||
DA448931.1 | - | 4, 5 | 203 | |||
DA483023.1 | - | 5, 6 | 250 | |||
Z70222.1 | - | 5, 6 | 221 |