Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDGFRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006206.4 | NP_006197.1 | 16 | 2635 | ||
XM_005265743.1 | XP_005265800.1 | 16 | 2419 | |||
XM_006714039.3 | XP_006714102.1 | 17 | 2680 | |||
XM_006714041.3 | XP_006714104.1 | 17 | 2683 | |||
XM_011534385.2 | XP_011532687.1 | 16 | 2673 | |||
XM_017008280.1 | XP_016863769.1 | 16 | 2850 | |||
XM_017008281.1 | XP_016863770.1 | 16 | 2916 | |||
XM_017008282.1 | XP_016863771.1 | 16 | 2564 | |||
GenBank mRNA | AK311006.1 | - | 15 | 2122 | ||
AK316578.1 | - | 16 | 2437 | |||
BC063414.1 | - | 17 | 2572 | 5205969 | ||
L25829.1 | - | 4 | 729 | |||
M21574.1 | - | 16 | 2443 | |||
M22734.1 | - | 2432 | ||||
X76079.1 | - | 4 | 729 |