Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260490.1 | NP_001247419.1 | 4, 5 | 1043 | ||
NM_015925.6 | NP_057009.3 | 6, 7 | 1310 | |||
NM_205834.3 | NP_991403.1 | 7, 8 | 1367 | |||
NM_205835.3 | NP_991404.1 | 5, 6 | 1163 | |||
XM_011527026.1 | XP_011525328.1 | 6, 7 | 1195 | |||
GenBank mRNA | AK075426.1 | - | 6, 7 | 962 | ||
AK130367.1 | - | 5, 6 | 817 | |||
AK296618.1 | - | 4, 5 | 697 | |||
BC000015.2 | - | 6, 7 | 941 | |||
BC004381.1 | - | 6, 7 | 958 | |||
BC047376.1 | - | 7, 8 | 1303 | 5272128 |