Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 2 | 533 | ||
NM_014955.2 | NP_055770.1 | 2 | 208 | |||
NM_015935.4 | NP_057019.3 | 2 | 533 | |||
XM_017001455.1 | XP_016856944.1 | 1 | 27 | |||
XM_017001456.1 | XP_016856945.1 | 1 | 27 | |||
XM_017001457.1 | XP_016856946.1 | 1 | 27 | |||
XM_017001458.1 | XP_016856947.1 | 2 | 650 | |||
XM_017001459.1 | XP_016856948.1 | 2 | 200 | |||
GenBank mRNA | AB020666.2 | - | 2 | 288 | ||
AB242174.1 | - | 2 | 458 | |||
AB242175.1 | - | 2 | 446 | |||
AF132936.1 | - | 2 | 458 | |||
AK027552.1 | - | 2 | 452 | |||
AK027621.1 | - | 2 | 147 | |||
AK223493.1 | - | 2 | 494 | |||
AK291740.1 | - | 2 | 480 | |||
AK304471.1 | - | 3 | 314 | |||
AL049669.1 | - | 2 | 458 | |||
BC029083.1 | - | 2 | 287 | 4509086 |