Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244585.1 | NP_001231514.1 | 5 | 443 | ||
NM_001270643.1 | NP_001257572.1 | 5 | 755 | |||
NM_016019.4 | NP_057103.2 | 4 | 684 | |||
GenBank mRNA | AF151817.1 | - | 4 | 243 | ||
AF151832.1 | - | 4 | 268 | |||
AK001476.1 | - | 4 | 576 | |||
AK001519.1 | - | 4 | 622 | |||
AK022895.1 | - | 4 | 559 | |||
AK092044.1 | - | 3 | 1065 | |||
AK093864.1 | - | 5 | 542 | |||
AK095839.1 | - | 3 | 1596 | |||
AK298166.1 | - | 5 | 443 | |||
BC017163.2 | - | 4 | 601 | 3920377 | ||
BC042625.1 | - | 5 | 689 | 5178074 | ||
BC050708.2 | - | 4 | 614 | 6500791 | ||
BC056886.1 | - | 4 | 559 | 6147116 |
Gene Symbol: C7orf55-LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244584.2 | NP_001231513.1 | 5 | 719 | ||
GenBank mRNA | AK297642.1 | - | 5 | 491 |