Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127399.1 | NP_001120871.1 | 4 | 534 | ||
NM_001127400.1 | NP_001120872.1 | 4 | 770 | |||
NM_001127401.1 | NP_001120873.1 | 5 | 831 | |||
NM_016061.2 | NP_057145.1 | 3 | 473 | |||
XM_017004318.1 | XP_016859807.1 | 5 | 1069 | |||
XM_017004319.1 | XP_016859808.1 | 4 | 507 | |||
XM_017004320.1 | XP_016859809.1 | 4 | 1008 | |||
XM_017004321.1 | XP_016859810.1 | 4 | 700 | |||
XM_017004322.1 | XP_016859811.1 | 4 | 931 | |||
GenBank mRNA | AB098739.1 | - | 2 | 270 | ||
AF135161.1 | - | 3 | 377 | |||
AF151885.1 | - | 3 | 395 | |||
AF172940.1 | - | 3 | 397 | |||
AK025730.1 | - | 4 | 411 | |||
AK307099.1 | - | 2 | 186 | |||
AK311404.1 | - | 3 | 184 | |||
BC000836.1 | - | 4 | 348 | 3456031 | ||
BC047237.1 | - | 4 | 931 | 5269563 |