Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316935.1 | NP_001303864.1 | 6 | 500 | ||
NM_016126.3 | NP_057210.2 | 6 | 697 | |||
NR_133632.1 | 4 | 521 | ||||
NR_133633.1 | 7 | 795 | ||||
GenBank mRNA | AF100747.1 | - | 6 | 469 | ||
AF416725.1 | - | 5 | 424 | |||
AF417114.1 | - | 5 | 424 | |||
AK225916.1 | - | 5 | 2584 | |||
AW024033.1 | - | 3 | 271 | |||
BC005245.1 | - | 6 | 489 | 3838843 | ||
BC045699.1 | - | 1 | 2118 |