Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016199.2 | NP_057283.1 | 2 | 115 | ||
XM_017026869.1 | XP_016882358.1 | 2 | 87 | |||
GenBank mRNA | AF182293.1 | - | 1 | 75 | ||
BC018621.1 | - | 1 | 68 | 4855162 | ||
BI553911.1 | - | 2 | 120 |