Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016200.4 | NP_057284.1 | 3 | 359 | ||
GenBank mRNA | AF182294.1 | - | 3 | 249 | ||
AK090561.1 | - | 1 | 1566 | |||
AK311936.1 | - | 3 | 275 | |||
BC002742.2 | - | 3 | 205 | 3626875 | ||
BC022440.1 | - | 3 | 229 | 4283482 | ||
DA112078.1 | - | 3 | 228 | |||
DA318705.1 | - | 3 | 326 | |||
DA478457.1 | - | 3 | 227 | |||
DB275674.1 | - | 3 | 360 |