Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302826.1 | NP_001289755.1 | 5 | 908 | ||
NM_001302827.1 | NP_001289756.1 | 4 | 382 | |||
NM_016229.4 | NP_057313.2 | 5 | 562 | |||
NR_126508.1 | 5 | 562 | ||||
XM_005252975.4 | XP_005253032.1 | 5 | 514 | |||
XM_006718251.2 | XP_006718314.1 | 5 | 539 | |||
XM_011520184.2 | XP_011518486.1 | 5 | 729 | |||
XM_011520185.2 | XP_011518487.1 | 7 | 828 | |||
XM_017017920.1 | XP_016873409.1 | 7 | 822 | |||
XM_017017921.1 | XP_016873410.1 | 7 | 824 | |||
XM_017017922.1 | XP_016873411.1 | 5 | 456 | |||
GenBank mRNA | AB209000.1 | - | 2 | 3955 | ||
AF169802.1 | - | 5 | 465 | |||
AK022556.1 | - | 1 | 1578 | |||
AK290102.1 | - | 5 | 881 | |||
AK308543.1 | - | 6 | 422 | |||
AL133582.1 | - | 4 | 298 | |||
AY665398.1 | - | 4 | 316 | |||
BC001346.1 | - | 5 | 436 | 3451742 | ||
BG723242.1 | - | 5 | 559 | |||
BM693762.1 | - | 5 | 490 | |||
BQ431043.1 | - | 4 | 320 | |||
CB149369.1 | - | 5 | 428 |