Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016243.2 | NP_057327.2 | 8 | 781 | ||
GenBank mRNA | AF087912.1 | - | 8 | 748 | ||
AF091084.1 | - | 3 | 206 | |||
AF093822.1 | - | 8 | 748 | |||
AF125533.1 | - | 8 | 748 | |||
AF169481.1 | - | 8 | 736 | |||
AK074654.1 | - | 8 | 762 | |||
AK075304.1 | - | 8 | 730 | |||
AK123705.1 | - | 2 | 1808 | |||
AK223416.1 | - | 8 | 762 | |||
AK313333.1 | - | 8 | 760 | |||
AY359026.1 | - | 8 | 785 | |||
BC018732.1 | - | 8 | 738 | 4638088 | ||
BC127945.1 | - | 5 | 468 | 40132732 |