Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291997.1 | NP_001278926.1 | 8 | 1416 | ||
NM_016373.3 | NP_057457.1 | 9 | 1481 | |||
XM_011523103.2 | XP_011521405.1 | 10 | 1316 | |||
XM_017023279.1 | XP_016878768.1 | 2 | 946 | |||
GenBank mRNA | AF211943.1 | - | 9 | 1240 | ||
AF227527.1 | - | 9 | 1219 | |||
AF395123.1 | - | 6 | 708 | |||
AF395124.1 | - | 5 | 601 | |||
AK027626.1 | - | 9 | 1239 | |||
AK290438.1 | - | 9 | 1231 | |||
AK291300.1 | - | 9 | 1238 | |||
AK298322.1 | - | 8 | 1203 | |||
AY256821.1 | - | 9 | 1168 | |||
BC003184.1 | - | 6 | 654 | 3535809 | ||
BT007445.1 | - | 575 |