Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016520.2 | NP_057604.1 | 2 | 174 | ||
GenBank mRNA | AF218421.1 | - | 2 | 129 | ||
BC007664.2 | - | 2 | 127 | 3140172 | ||
BC017570.1 | - | 2 | 131 | 4109447 | ||
CB128496.1 | - | 2 | 174 |