Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042452.1 | NP_001035917.1 | 4 | 372 | ||
NM_001042453.1 | NP_001035918.1 | 4 | 432 | |||
NM_016542.3 | NP_057626.2 | 5 | 663 | |||
XM_011531349.1 | XP_011529651.1 | 6 | 636 | |||
XM_011531350.1 | XP_011529652.1 | 5 | 709 | |||
GenBank mRNA | AB040057.2 | - | 5 | 582 | ||
AF231012.1 | - | 5 | 551 | |||
AF344882.1 | - | 4 | 372 | |||
AF344883.1 | - | 4 | 372 | |||
AK075107.1 | - | 4 | 344 | |||
AK075150.1 | - | 5 | 578 | |||
AK303583.1 | - | 6 | 644 | |||
AK314356.1 | - | 5 | 579 | |||
AK316137.1 | - | 6 | 644 | |||
BC098315.1 | - | 5 | 568 | 40026349 | ||
BC099843.1 | - | 5 | 568 | 40026348 | ||
BC103503.1 | - | 5 | 568 | 40026350 | ||
BT020099.1 | - | 4 | 362 | |||
DA760264.1 | - | 4 | 436 |