Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207012.1 | NP_001193941.1 | 4 | 875 | ||
NM_001293643.1 | NP_001280572.1 | 4 | 872 | |||
NM_002632.5 | NP_002623.2 | 4 | 875 | |||
GenBank mRNA | A18411.1 | - | 4 | 674 | ||
AK309886.1 | - | 4 | 708 | |||
AK310608.1 | - | 4 | 708 | |||
AK310886.1 | - | 3 | 666 | |||
AL543693.3 | - | 4 | 686 | |||
BC001422.2 | - | 4 | 699 | 3139175 | ||
BC007255.1 | - | 4 | 680 | 3028008 | ||
BC007789.2 | - | 4 | 677 | 3629125 | ||
BT007182.1 | - | 4 | 353 | |||
BX248289.1 | - | 4 | 875 | |||
S72960.1 | - | 4 | 365 | |||
X54936.1 | - | 4 | 674 |