Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293643.1 | NP_001280572.1 | 5, 6 | 930 | ||
NM_002632.5 | NP_002623.2 | 5, 6 | 933 | |||
GenBank mRNA | AK023843.1 | - | 1, 2 | 1825 | ||
AL543693.3 | - | 5, 6 | 744 | |||
BC001422.2 | - | 5, 6 | 757 | 3139175 | ||
BC007255.1 | - | 5, 6 | 738 | 3028008 | ||
BC007789.2 | - | 5, 6 | 735 | 3629125 | ||
BT007182.1 | - | 5, 6 | 411 | |||
BX248289.1 | - | 5, 6 | 933 | |||
S72960.1 | - | 5, 6 | 423 |