Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGGT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005023.3 | NP_005014.2 | 4 | 438 | ||
XM_005272020.3 | XP_005272077.2 | 4 | 453 | |||
XM_011543490.2 | XP_011541792.1 | 4 | 453 | |||
XM_011543492.2 | XP_011541794.1 | 4 | 450 | |||
GenBank mRNA | AY780790.1 | - | 4 | 417 | ||
DA024020.1 | - | 4 | 438 |