Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199917.1 | NP_001186846.1 | 5, 6 | 711 | ||
NM_001199918.1 | NP_001186847.1 | 3, 4 | 369 | |||
NM_001199919.1 | NP_001186848.1 | 4, 5 | 575 | |||
NM_015599.2 | NP_056414.1 | 4, 5 | 575 | |||
XM_017010935.1 | XP_016866424.1 | 3, 4 | 342 | |||
XM_017010936.1 | XP_016866425.1 | 4, 5 | 574 | |||
XM_017010937.1 | XP_016866426.1 | 3, 4 | 342 | |||
XM_017010938.1 | XP_016866427.1 | 4, 5 | 563 | |||
GenBank mRNA | AB032081.1 | - | 4, 5 | 487 | ||
AF102265.1 | - | 4, 5 | 491 | |||
AF180371.1 | - | 4, 5 | 498 | |||
AK001365.1 | - | 3, 4 | 310 | |||
AK023432.1 | - | 4, 5 | 520 | |||
AK225363.1 | - | 4, 5 | 513 | |||
AK301867.1 | - | 5, 6 | 689 | |||
AK314512.1 | - | 4, 5 | 508 | |||
BC001258.1 | - | 4, 5 | 534 | 3451677 |