Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002641.3 | NP_002632.1 | 4 | 1074 | ||
NM_020473.3 | NP_065206.3 | 3 | 297 | |||
NR_033835.1 | 4 | 700 | ||||
NR_033836.1 | 4 | 416 | ||||
XM_011545539.2 | XP_011543841.1 | 3 | 1898 | |||
XM_017029581.1 | XP_016885070.1 | 3 | 1021 | |||
GenBank mRNA | AK098047.1 | - | 3 | 510 | ||
AK303538.1 | - | 3 | 289 | |||
AK304144.1 | - | 4 | 1043 | |||
AK315063.1 | - | 3 | 1033 | |||
BC038236.1 | - | 3 | 1031 | 5733899 | ||
CX870731.1 | - | 3 | 376 | |||
D11466.1 | - | 4 | 1043 | |||
DA951065.1 | - | 4 | 416 | |||
S61523.1 | - | 1 | 58 | |||
S74936.1 | - | 2 | 516 |