Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005027.3 | NP_005018.1 | 11, 12 | 1956 | ||
GenBank mRNA | BC011917.2 | - | 3, 4 | 398 | ||
BC014170.2 | - | 2, 3 | 206 | |||
BC070082.1 | - | 11, 12 | 1928 | 30343107 | ||
BC090249.1 | - | 11, 12 | 1888 | 6341766 | ||
X80907.1 | - | 10, 11 | 1657 |