Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206796.2 | NP_001193725.1 | 11, 12 | 2358 | ||
NM_001206797.2 | NP_001193726.1 | 9, 10 | 1799 | |||
NM_001206798.2 | NP_001193727.1 | 10, 11 | 2046 | |||
NM_001206799.1 | NP_001193728.1 | 10, 11 | 1688 | |||
NM_001316318.1 | NP_001303247.1 | 10, 11 | 2241 | |||
NM_002654.5 | NP_002645.3 | 10, 11 | 2021 | |||
NM_182470.3 | NP_872270.1 | 10, 11 | 2237 | |||
NM_182471.3 | NP_872271.1 | 10, 11 | 2021 | |||
XM_005254443.1 | XP_005254500.1 | 11, 12 | 1745 | |||
XM_005254445.4 | XP_005254502.1 | 10, 11 | 1582 | |||
XM_006720570.1 | XP_006720633.1 | 11, 12 | 1842 | |||
XM_011521670.2 | XP_011519972.1 | 10, 11 | 1807 | |||
XM_017022313.1 | XP_016877802.1 | 10, 11 | 1582 | |||
GenBank mRNA | AF025439.1 | - | 3, 4 | 398 | ||
AK092369.1 | - | 10, 11 | 1771 | |||
AK222927.1 | - | 10, 11 | 1587 | |||
AK294315.1 | - | 11, 12 | 1847 | |||
AK297951.1 | - | 1365 | ||||
AK299413.1 | - | 10, 11 | 1525 | |||
AK300800.1 | - | 10, 11 | 1612 | |||
AK307855.1 | - | 11, 12 | 1589 | |||
AK309781.1 | - | 10, 11 | 1690 | |||
AK312253.1 | - | 10, 11 | 1587 | |||
BC000481.2 | - | 10, 11 | 1603 | 2964687 | ||
BC007640.1 | - | 10, 11 | 1587 | 3533876 | ||
BC007952.2 | - | 10, 11 | 1599 | 4299213 | ||
BC012811.2 | - | 10, 11 | 1599 | 2958817 | ||
BC019265.2 | - | 5, 6 | 935 | |||
BC023328.1 | - | 6, 7 | 971 | |||
BC023592.2 | - | 10, 11 | 1549 | |||
BC035198.1 | - | 10, 11 | 1553 | 3859987 | ||
BC094767.1 | - | 7, 8 | 1148 | 5296639 | ||
BC096823.1 | - | 10, 11 | 1788 | |||
BX648597.1 | - | 1, 2 | 1590 | |||
EU794631.1 | - | 10, 11 | 1740 | |||
M23725.1 | - | 10, 11 | 1608 | |||
M26252.1 | - | 10, 11 | 1588 |