Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000928.2 | NP_000919.1 | 2 | 164 | ||
GenBank mRNA | AA844927.1 | - | 3 | 404 | ||
AK311830.1 | - | 2 | 149 | |||
BC005386.1 | - | 2 | 154 | |||
BC106725.1 | - | 2 | 164 | 40014551 | ||
BC106726.1 | - | 2 | 164 | 40014552 | ||
BE969737.1 | - | 135 | ||||
M21054.1 | - | 2 | 165 |