Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000928.2 | NP_000919.1 | 4 | 374 | ||
GenBank mRNA | AA844927.1 | - | 1 | 194 | ||
AK311830.1 | - | 4 | 359 | |||
BC005386.1 | - | 3 | 236 | |||
BC106725.1 | - | 4 | 374 | 40014551 | ||
BC106726.1 | - | 4 | 374 | 40014552 | ||
BE969737.1 | - | 345 | ||||
BX113838.1 | - | 3 | 178 | |||
CA867923.1 | - | 3 | 217 | |||
M21054.1 | - | 4 | 375 |