Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278603.1 | NP_001265532.1 | 6 | 943 | ||
NM_017426.3 | NP_059122.2 | 7 | 1087 | |||
NR_103780.1 | 6 | 860 | ||||
NR_103781.1 | 6 | 860 | ||||
NR_103782.1 | 7 | 857 | ||||
XM_006714237.1 | XP_006714300.1 | 8 | 1064 | |||
XM_011532033.2 | XP_011530335.1 | 7 | 1030 | |||
XM_011532034.2 | XP_011530336.1 | 6 | 1330 | |||
XM_017008294.1 | XP_016863783.1 | 7 | 1474 | |||
XM_017008295.1 | XP_016863784.1 | 6 | 1247 | |||
GenBank mRNA | AF157322.1 | - | 7 | 965 | ||
AK001517.1 | - | 7 | 969 | |||
AK222752.1 | - | 7 | 974 | |||
AK223352.1 | - | 7 | 970 | |||
AK299442.1 | - | 6 | 742 | |||
AK300036.1 | - | 6 | 825 | |||
AK307571.1 | - | 7 | 747 | |||
AK315160.1 | - | 7 | 967 | |||
BC012559.1 | - | 7 | 984 | |||
BX538002.1 | - | 6 | 760 |