Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000301.3 | NP_000292.1 | 8 | 941 | ||
GenBank mRNA | AK312966.1 | - | 8 | 892 | ||
BC060513.1 | - | 8 | 867 | 4609795 | ||
M74220.1 | - | 8 | 878 | |||
X05199.1 | - | 8 | 883 |