Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002668.2 | NP_002659.1 | 1 | 236 | ||
GenBank mRNA | BC109066.2 | - | 1 | 137 | 40033888 | |
DB220888.1 | - | 1 | 237 | |||
L09604.1 | - | 1 | 147 |