Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242920.1 | NP_001229849.1 | 10, 11 | 1187 | ||
NM_001242921.1 | NP_001229850.1 | 10, 11 | 1099 | |||
NM_006227.3 | NP_006218.1 | 12, 13 | 1472 | |||
NM_182676.2 | NP_872617.1 | 11, 12 | 1316 | |||
GenBank mRNA | AB076694.1 | - | 12, 13 | 1256 | ||
AF052157.1 | - | 9, 10 | 974 | |||
AK092397.1 | - | 12, 13 | 1213 | |||
AK092523.1 | - | 11, 12 | 1104 | |||
AK097009.1 | - | 11, 12 | 1770 | |||
AK222690.1 | - | 12, 13 | 1260 | |||
AK289371.1 | - | 11, 12 | 1104 | |||
AK293150.1 | - | 10, 11 | 1015 | |||
AK299181.1 | - | 10, 11 | 979 | |||
BC005045.1 | - | 11, 12 | 1097 | 3510757 | ||
BC019847.1 | - | 12, 13 | 1254 | 4997011 | ||
BC019898.1 | - | 12, 13 | 1247 | 4992839 | ||
L26232.1 | - | 12, 13 | 1262 |