Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144717.3 | NP_653318.2 | 3, 4 | 663 | ||
XM_006713665.3 | XP_006713728.1 | 3, 4 | 1392 | |||
XM_011512910.2 | XP_011511212.1 | 4, 5 | 567 | |||
XM_011512911.2 | XP_011511213.1 | 3, 4 | 520 | |||
XM_017006632.1 | XP_016862121.1 | 2, 3 | 476 | |||
GenBank mRNA | AK296832.1 | - | 3, 4 | 535 | ||
AK316084.1 | - | 3, 4 | 543 | |||
AY358305.1 | - | 3, 4 | 663 | |||
BC033292.1 | - | 3, 4 | 654 | |||
BC063696.1 | - | 3, 4 | 527 | 4746920 |