Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023915.3 | NP_076404.3 | 3 | 1171 | ||
GenBank mRNA | AB031548.1 | - | 3 | 1103 | ||
AF086432.1 | - | 1 | 246 | |||
AF237763.1 | - | 3 | 1059 | |||
AF345566.1 | - | 1 | 849 | |||
AF411114.1 | - | 2 | 810 | |||
AK027784.1 | - | 3 | 1171 | |||
BC009540.1 | - | 1 | 617 | |||
BC112941.1 | - | 3 | 1080 | 4500655 | ||
BC117565.1 | - | 3 | 1080 | 4521950 | ||
BC132888.1 | - | 3 | 1137 | 40146910 | ||
BC132890.1 | - | 3 | 1137 | 40146912 | ||
EU883576.1 | - | 2 | 810 |
Set Membership: |
G-protein coupled receptor |