Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 9, 10 | 1069 | ||
NM_001322003.1 | NP_001308932.1 | 9, 10 | 1064 | |||
NM_001322004.1 | NP_001308933.1 | 8, 9 | 929 | |||
NM_001322005.1 | NP_001308934.1 | 9, 10 | 1181 | |||
NM_001322007.1 | NP_001308936.1 | 8, 9 | 966 | |||
NM_001322008.1 | NP_001308937.1 | 7, 8 | 826 | |||
NM_001322009.1 | NP_001308938.1 | 9, 10 | 1176 | |||
NM_001322011.1 | NP_001308940.1 | 9, 10 | 1020 | |||
NM_001322012.1 | NP_001308941.1 | 9, 10 | 1015 | |||
NM_001322013.1 | NP_001308942.1 | 8, 9 | 896 | |||
NM_001322014.1 | NP_001308943.1 | 9, 10 | 1069 | |||
NM_001322015.1 | NP_001308944.1 | 9, 10 | 1239 | |||
NR_136154.1 | 9, 10 | 1069 | ||||
XM_006715744.3 | XP_006715807.1 | 2, 3 | 145 | |||
XM_017012342.1 | XP_016867831.1 | 8, 9 | 894 | |||
GenBank mRNA | AB103082.1 | - | 9, 10 | 982 | ||
AB103086.1 | - | 6, 7 | 664 | |||
AK294661.1 | - | 8, 9 | 919 | |||
AK312390.1 | - | 9, 10 | 1018 | |||
BC008400.1 | - | 9, 10 | 1027 | |||
BC031832.1 | - | 9, 10 | 1033 | |||
BC093921.1 | - | 9, 10 | 1004 | 7939766 | ||
BP251389.1 | - | 5, 6 | 539 | |||
U14658.1 | - | 9, 10 | 984 |