Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 8, 9 | 993 | ||
NM_001322003.1 | NP_001308932.1 | 8, 9 | 988 | |||
NM_001322004.1 | NP_001308933.1 | 7, 8 | 853 | |||
NM_001322005.1 | NP_001308934.1 | 8, 9 | 1105 | |||
NM_001322006.1 | NP_001308935.1 | 8, 9 | 993 | |||
NM_001322007.1 | NP_001308936.1 | 7, 8 | 890 | |||
NM_001322008.1 | NP_001308937.1 | 6, 7 | 750 | |||
NM_001322009.1 | NP_001308938.1 | 8, 9 | 1100 | |||
NM_001322010.1 | NP_001308939.1 | 7, 8 | 853 | |||
NM_001322011.1 | NP_001308940.1 | 8, 9 | 944 | |||
NM_001322012.1 | NP_001308941.1 | 8, 9 | 939 | |||
NM_001322013.1 | NP_001308942.1 | 7, 8 | 820 | |||
NM_001322014.1 | NP_001308943.1 | 8, 9 | 993 | |||
NM_001322015.1 | NP_001308944.1 | 8, 9 | 1163 | |||
NR_136154.1 | 8, 9 | 993 | ||||
GenBank mRNA | AB103082.1 | - | 8, 9 | 906 | ||
AB103086.1 | - | 5, 6 | 588 | |||
AK294661.1 | - | 7, 8 | 843 | |||
AK312390.1 | - | 8, 9 | 942 | |||
BC008400.1 | - | 8, 9 | 951 | |||
BC031832.1 | - | 8, 9 | 957 | |||
BC093921.1 | - | 8, 9 | 928 | 7939766 | ||
BP251389.1 | - | 4, 5 | 463 | |||
U14658.1 | - | 8, 9 | 908 |