Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322270.1 | NP_001309199.1 | 14, 15 | 2375 | ||
NM_001322271.1 | NP_001309200.1 | 14, 15 | 2368 | |||
NM_001322272.1 | NP_001309201.1 | 15, 16 | 2733 | |||
NM_017437.2 | NP_059133.1 | 15, 16 | 2528 | |||
GenBank mRNA | AB037788.1 | - | 9, 10 | 1652 | ||
AK001627.2 | - | 15, 16 | 2505 | |||
AK001912.1 | - | 13, 14 | 2084 | |||
AK023583.1 | - | 12, 13 | 2105 | |||
AL442079.1 | - | 8, 9 | 1427 | |||
BC070095.1 | - | 15, 16 | 2498 | 5267724 |