Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242590.2 | NP_001229519.1 | 3 | 512 | ||
NM_018643.4 | NP_061113.1 | 4 | 705 | |||
NR_136332.1 | 4 | 705 | ||||
XM_017010956.1 | XP_016866445.1 | 4 | 645 | |||
XM_017010957.1 | XP_016866446.1 | 3 | 449 | |||
GenBank mRNA | AF196329.1 | - | 4 | 662 | ||
AF287008.1 | - | 4 | 679 | |||
AK292162.1 | - | 4 | 642 | |||
AK301519.1 | - | 3 | 449 | |||
AK313104.1 | - | 4 | 657 | |||
AY074783.1 | - | 3 | 439 | |||
BC017773.1 | - | 4 | 660 | 4692680 | ||
DA362458.1 | - | 3 | 152 |