Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040875.2 | NP_001035808.1 | 4, 5 | 531 | ||
NM_018972.2 | NP_061845.2 | 4, 5 | 661 | |||
NR_046346.1 | 4, 5 | 516 | ||||
XM_011517551.2 | XP_011515853.1 | 3, 4 | 1619 | |||
XM_011517552.2 | XP_011515854.1 | 3, 4 | 456 | |||
XM_017013585.1 | XP_016869074.1 | 4, 5 | 1793 | |||
GenBank mRNA | AB551556.1 | - | 4, 5 | 660 | ||
AB551557.1 | - | 4, 5 | 660 | |||
AB551558.1 | - | 4, 5 | 658 | |||
AK292572.1 | - | 4, 5 | 600 | |||
AK295594.1 | - | 4, 5 | 455 | |||
BC024939.1 | - | 4, 5 | 496 | 3923335 | ||
BC035450.1 | - | 4, 5 | 508 | |||
Y17849.1 | - | 4, 5 | 589 |