Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271581.1 | NP_001258510.1 | 4 | 984 | ||
NM_018981.2 | NP_061854.1 | 4 | 984 | |||
NR_073365.1 | 4 | 984 | ||||
NR_073366.1 | 4 | 984 | ||||
NR_073367.1 | 4 | 984 | ||||
GenBank mRNA | AF038503.1 | - | 4 | 695 | ||
AF314529.1 | - | 2 | 338 | |||
AF490904.1 | - | 4 | 732 | |||
AK027450.1 | - | 2 | 325 | |||
AK027647.1 | - | 4 | 753 | |||
AK027696.1 | - | 4 | 732 | |||
AK027701.2 | - | 1 | 106 | |||
AL832632.2 | - | 4 | 755 | |||
AL832646.3 | - | 2 | 336 | |||
AY089971.1 | - | 2 | 338 | |||
AY358577.1 | - | 4 | 741 | |||
BC070034.1 | - | 4 | 729 | |||
BC107425.1 | - | 2 | 361 | |||
BC117299.1 | - | 4 | 699 | 40125850 | ||
BC126168.1 | - | 4 | 699 | 8991884 |