Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018984.3 | NP_061857.3 | 15 | 2526 | ||
XM_005268984.4 | XP_005269041.1 | 15 | 3237 | |||
XM_005268985.2 | XP_005269042.1 | 14 | 2352 | |||
XM_011538497.1 | XP_011536799.1 | 14 | 2618 | |||
XM_011538499.1 | XP_011536801.1 | 13 | 2417 | |||
XM_011538500.2 | XP_011536802.1 | 14 | 2442 | |||
XM_011538501.2 | XP_011536803.1 | 14 | 2438 | |||
XM_017019491.1 | XP_016874980.1 | 14 | 3464 | |||
XM_017019492.1 | XP_016874981.1 | 15 | 2579 | |||
XM_017019493.1 | XP_016874982.1 | 13 | 2323 | |||
XM_017019494.1 | XP_016874983.1 | 13 | 2418 | |||
GenBank mRNA | AB037719.1 | - | 5 | 1561 | ||
AB072355.1 | - | 15 | 2439 | |||
BC062341.1 | - | 15 | 2518 | 6376684 |