Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3TC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318480.1 | NP_001305409.1 | 14 | 3500 | ||
NM_018986.4 | NP_061859.3 | 14 | 3417 | |||
NR_134639.1 | 12 | 3131 | ||||
XM_006713889.2 | XP_006713952.1 | 11 | 2835 | |||
XM_006713891.3 | XP_006713954.1 | 8 | 2477 | |||
XM_011513488.1 | XP_011511790.1 | 14 | 3394 | |||
XM_011513489.2 | XP_011511791.1 | 12 | 3017 | |||
XM_017008300.1 | XP_016863789.1 | 14 | 3526 | |||
XM_017008301.1 | XP_016863790.1 | 14 | 3209 | |||
XM_017008302.1 | XP_016863791.1 | 11 | 2919 | |||
GenBank mRNA | AK074093.1 | - | 13 | 4211 | ||
AK074402.1 | - | 14 | 3294 | |||
AK093562.1 | - | 3 | 1338 | |||
AK126165.1 | - | 14 | 3373 | |||
AK128258.1 | - | 2 | 1111 | |||
AK310552.1 | - | 3 | 358 |