Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018997.3 | NP_061870.1 | 2 | 748 | ||
NM_031901.5 | NP_114107.1 | 3 | 312 | |||
GenBank mRNA | AA977333.1 | - | 1 | 201 | ||
AB049955.1 | - | 3 | 285 | |||
AF182417.1 | - | 2 | 257 | |||
AK058101.1 | - | 2 | 914 | |||
AK289467.1 | - | 3 | 299 | |||
BC004566.2 | - | 3 | 281 | 3633098 | ||
BC006554.2 | - | 2 | 653 | 2819715 | ||
BI755132.1 | - | 2 | 705 | |||
HY161691.1 | - | 3 | 313 | |||
U79258.1 | - | 2 | 669 |