Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018998.3 | NP_061871.1 | 2 | 321 | ||
XM_005266089.2 | XP_005266146.2 | 2 | 341 | |||
XM_011518789.1 | XP_011517091.1 | 2 | 341 | |||
XM_017014812.1 | XP_016870301.1 | 2 | 341 | |||
GenBank mRNA | AK026081.1 | - | 2 | 185 | ||
AK315738.1 | - | 2 | 196 | |||
BC014130.1 | - | 2 | 217 | 4620371 | ||
BC014297.2 | - | 2 | 188 | 3958395 | ||
BC031100.1 | - | 2 | 263 | |||
BC035355.1 | - | 2 | 255 | |||
BI829307.1 | - | 2 | 288 | |||
CF132404.1 | - | 2 | 262 | |||
DB460097.1 | - | 2 | 305 |